Läs mer
2,3,4,5-Tetrathiahexane, TRC
Produktkod. 30418351
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30418351 100 mg
1 options
Produktkod. 30418351 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT011235100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2,3,4,5-Tetrathiahexane
CAS 5756-24-1
Molekylformel C2 H6 S4
Kvantitet 100 mg
Synonym 1,4-Dimethyltetrasulfane, (Methyltetrasulfanyl)methane, Dimethyl Tetrasulfide, Methyl tetrasulfide
InChI formel InChI=1S/C2H6S4/c1-3-5-6-4-2/h1-2H3
LEDER CSSSSC
IUPAC-namn (methyltetrasulfanyl)methane
Molekylvikt (g/mol) 158.329
Formel vikt 157.935