Läs mer
2,3,4,6-Tetra-O-pivaloyl-Beta-D-galactopyranosyl Isothiocyanate, TRC
Produktkod. 30449065
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30449065 10 mg
1 options
Produktkod. 30449065 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT30636510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2,3,4,6-Tetra-O-pivaloyl-beta-D-galactopyranosyl Isothiocyanate
CAS 147948-52-5
Molekylformel C27H43NO9S
Kvantitet 10 mg
Synonym beta-D-Galactopyranosyl isothiocyanate 2,3,4,6-tetrakis(2,2-dimethylpropanoate), 2,3,4,6-Tetra-O-pivaloyl-b-D-galactopyranosyl Isothiocyanate
InChI formel InChI=1S/C27H43NO9S/c1-24(2,3)20(29)33-13-15-16(35-21(30)25(4,5)6)17(36-22(31)26(7,8)9)18(19(34-15)28-14-38)37-23(32)27(10,11)12/h15-19H,13H2,1-12H3/t15-,16+,17+,18-,19-/m1/s1
LEDER CC(C)(C)C(=O)OC[C@H]1O[C@@H](N=C=S)[C@H](OC(=O)C(C)(C)C)[C@@H](OC(=O)C(C)(C)C)[C@H]1OC(=O)C(C)(C)C
IUPAC-namn [(2R,3S,4S,5R,6R)-3,4,5-tris(2,2-dimethylpropanoyloxy)-6-isothiocyanatooxan-2-yl]methyl 2,2-dimethylpropanoate
Molekylvikt (g/mol) 557.7
Formel vikt 557.2659