Läs mer
2-[[3,4,6-Tri-O-acetyl-2-(acetylamino)-2-deoxy-Beta-D-galactopyranosyl]oxy]acetic Acid, TRC
Produktkod. 30452267
Change view
Kvantitet:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30452267 500 mg
30451340 50 mg
30453476 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30452267 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT992240500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1159409-18-3
Molekylformel C16H23NO11
Molekylvikt (g/mol) 405.35
Synonym 2-(((2R,3R,4R,5R,6R)-3-Acetamido-4,5-diacetoxy-6-(acetoxymethyl)tetrahydro-2H-pyran-2-yl)oxy)acetic Acid
IUPAC-namn 2-[(2R,3R,4R,5R,6R)-3-acetamido-4,5-diacetyloxy-6-(acetyloxymethyl)oxan-2-yl]oxyacetic acid
LEDER CC(=O)N[C@H]1[C@H](OCC(=O)O)O[C@H](COC(=O)C)[C@H](OC(=O)C)[C@@H]1OC(=O)C
Specifikationer
Kemiskt namn eller material 2-((3,4,6-Tri-O-acetyl-2-(acetylamino)-2-deoxy-beta-D-galactopyranosyl)oxy)acetic Acid
Kvantitet 500 mg
InChI formel InChI=1S/C16H23NO11/c1-7(18)17-13-15(27-10(4)21)14(26-9(3)20)11(5-24-8(2)19)28-16(13)25-6-12(22)23/h11,13-16H,5-6H2,1-4H3,(H,17,18)(H,22,23)/t11-,13-,14+,15-,16-/m1/s1
Formel vikt 405.1271