missing translation for 'onlineSavingsMsg'
Läs mer

α,α'-[[[2-(3,4-Dimethoxyphenyl)ethyl]imino]di-3,1-propanediyl]bis[3,4-dimethoxy-α-(1-methylethyl)-benzeneacetonitrile, TRC

Produktkod. 30374884
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
2.5 mg
Förpackningsstorlek:
2.5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30374884 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30374884 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4599352.5MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material 5,5'-((2-(3,4-Dimethoxyphenyl)ethyl)imino)bis(2-(3,4-dimethoxyphenyl)-2-(1-methylethyl)-pentanenitrile)
CAS 141991-89-1
Molekylformel C42 H57 N3 O6
Kvantitet 2.5 mg
Synonym Benzeneacetonitrile, α,α'-[[[2-(3,4-dimethoxyphenyl)ethyl]imino]di-3,1-propanediyl]bis[3,4-dimethoxy-α-(1-methylethyl)-, α,α'-[[[2-(3,4-Dimethoxyphenyl)ethyl]imino]di-3,1-propanediyl]bis[3,4-dimethoxy-α-(1-methylethyl)benzeneacetonitrile], 5,5'-[[2-(3,4-D
InChI formel InChI=1S/C42H57N3O6/c1-30(2)41(28-43,33-14-17-36(47-6)39(26-33)50-9)20-11-22-45(24-19-32-13-16-35(46-5)38(25-32)49-8)23-12-21-42(29-44,31(3)4)34-15-18-37(48-7)40(27-34)51-10/h13-18,25-27,30-31H,11-12,19-24H2,1-10H3
LEDER COc1ccc(CCN(CCCC(C#N)(C(C)C)c2ccc(OC)c(OC)c2)CCCC(C#N)(C(C)C)c3ccc(OC)c(OC)c3)cc1OC
IUPAC-namn 5-[[4-cyano-4-(3,4-dimethoxyphenyl)-5-methylhexyl]-[2-(3,4-dimethoxyphenyl)ethyl]amino]-2-(3,4-dimethoxyphenyl)-2-propan-2-ylpentanenitrile
Molekylvikt (g/mol) 699.918
Formel vikt 699.425
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.