Läs mer
2,3,4-Tri-O-acetyl-a-D-glucuronic Acid Methyl Ester Trichloroacetimidate, TRC
Produktkod. 30452747
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30452747 1 g
1 options
Produktkod. 30452747 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT7652001G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2,3,4-Tri-O-acetyl-alpha-D-glucuronic acid methyl ester trichloroacetimidate
CAS 92420-89-8
Molekylformel C15 H18 Cl3 N O10
Kvantitet 1 g
Synonym 2,3,4-Tri-O-Acetyl-α-D-Glucuronide Methyl Ester Trichloroacetimidate
InChI formel InChI=1S/C15H18Cl3NO10/c1-5(20)25-8-9(26-6(2)21)11(27-7(3)22)13(28-10(8)12(23)24-4)29-14(19)15(16,17)18/h8-11,13,19H,1-4H3/t8-,9-,10-,11+,13+/m0/s1
LEDER COC(=O)[C@H]1O[C@H](OC(=N)C(Cl)(Cl)Cl)[C@H](OC(=O)C)[C@@H](OC(=O)C)[C@@H]1OC(=O)C
IUPAC-namn methyl (2S,3S,4S,5R,6R)-3,4,5-triacetyloxy-6-(2,2,2-trichloroethanimidoyl)oxyoxane-2-carboxylate
Molekylvikt (g/mol) 478.66
Formel vikt 476.9996