Läs mer
2,3,4-Tri-O-acetyl-D-glucuronic Acid Methyl Ester, TRC
Produktkod. 30453675
Change view
Kvantitet:
5 g
Produktkod. Kvantitet
30453675 5 g
1 options
Produktkod. 30453675 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT7651805G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2,3,4-Tri-O-acetyl-a-D-glucuronic Acid Methyl Ester
CAS 72692-06-9
Molekylformel C13 H18 O10
Kvantitet 5 g
Synonym 2,3,4-Tri-O-Acetyl-α-D-Glucuronic Acid Methyl Ester
InChI formel InChI=1S/C13H18O10/c1-5(14)20-8-9(21-6(2)15)11(22-7(3)16)13(18)23-10(8)12(17)19-4/h8-11,13,18H,1-4H3/t8-,9-,10-,11+,13-/m0/s1
LEDER COC(=O)[C@H]1O[C@H](O)[C@H](OC(=O)C)[C@@H](OC(=O)C)[C@@H]1OC(=O)C
IUPAC-namn methyl (2S,3S,4S,5R,6S)-3,4,5-triacetyloxy-6-hydroxyoxane-2-carboxylate
Molekylvikt (g/mol) 334.28
Formel vikt 334.09