Läs mer
2,3,4-Tri-O-acetyl-D-xylopyranose, TRC
Produktkod. 30450555
Change view
Kvantitet:
50 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30450555 50 mg
30451055 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30450555 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT76702050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2,3,4-Tri-O-acetyl-D-xylopyranose
CAS 55018-54-7
Molekylformel C11 H16 O8
Kvantitet 50 mg
InChI formel InChI=1S/C11H16O8/c1-5(12)17-8-4-16-11(15)10(19-7(3)14)9(8)18-6(2)13/h8-11,15H,4H2,1-3H3/t8-,9+,10-,11?/m1/s1
LEDER CC(=O)O[C@@H]1COC(O)[C@H](OC(=O)C)[C@H]1OC(=O)C
IUPAC-namn [(3R,4S,5R)-4,5-diacetyloxy-6-hydroxyoxan-3-yl] acetate
Molekylvikt (g/mol) 276.24
Formel vikt 276.0845