Läs mer
2,3,4,-Tri-O-benzyl-L-fucopyranose, TRC
Produktkod. 30455865
Change view
Kvantitet:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30455865 250 mg
30456737 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30455865 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT768000250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2,3,4,-Tri-O-benzyl-L-fucopyranose
CAS 60431-34-7
Molekylformel C27 H30 O5
Kvantitet 250 mg
InChI formel InChI=1S/C27H30O5/c1-20-24(29-17-21-11-5-2-6-12-21)25(30-18-22-13-7-3-8-14-22)26(27(28)32-20)31-19-23-15-9-4-10-16-23/h2-16,20,24-28H,17-19H2,1H3/t20-,24+,25+,26-,27?/m0/s1
LEDER C[C@@H]1OC(O)[C@@H](OCc2ccccc2)[C@H](OCc3ccccc3)[C@@H]1OCc4ccccc4
IUPAC-namn (3S,4R,5R,6S)-6-methyl-3,4,5-tris(phenylmethoxy)oxan-2-ol
Molekylvikt (g/mol) 434.52
Formel vikt 434.2093