Läs mer
2,3,4-Trichlorophenylboronic Acid, TRC
Produktkod. 30452544
Change view
Kvantitet:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30452544 50 mg
30450815 500 mg
30449679 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30452544 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT89624550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 352530-21-3
Molekylformel C6H4BCl3O2
Molekylvikt (g/mol) 225.26
IUPAC-namn (2,3,4-trichlorophenyl)boronic acid
LEDER OB(O)c1ccc(Cl)c(Cl)c1Cl
Specifikationer
Kemiskt namn eller material 2,3,4-Trichlorophenylboronic Acid
Kvantitet 50 mg
InChI formel InChI=1S/C6H4BCl3O2/c8-4-2-1-3(7(11)12)5(9)6(4)10/h1-2,11-12H
Formel vikt 223.937