Läs mer
2,3,4-Trichlorotoluene, TRC
Produktkod. 17983065
Change view
Kvantitet:
500 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17983065 500 mg
17903575 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17983065 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT774435500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 7359-72-0
Molekylformel C7H5Cl3
Molekylvikt (g/mol) 195.47
Synonym 1,2,3-Trichloro-4-methylbenzene (ACI), Toluene, 2,3,4-trichloro- (7CI, 8CI), 2,3,4-Trichlorotoluene, 2,3,4-Trichlorotoluene;
IUPAC-namn 1,2,3-trichloro-4-methylbenzene
LEDER Cc1ccc(Cl)c(Cl)c1Cl
Specifikationer
Kemiskt namn eller material 2,3,4-Trichlorotoluene
Kvantitet 500 mg
Anteckningar om renhetsgrad HPLC
InChI formel InChI=1S/C7H5Cl3/c1-4-2-3-5(8)7(10)6(4)9/h2-3H,1H3
Formel vikt 193.9457
Procent renhet >95%