missing translation for 'onlineSavingsMsg'
Learn More

2,3,4-Trihydroxyacetofenon, TRC

Product Code. 30450049
Change view
Click to view available options
Kvantitet:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30450049 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30450049 Supplier Toronto Research Chemicals Supplier No. TRCT79490510G
 Begränsad artikel
Beräknad leverans: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Högrena organiska molekyler och analytiska standarder, strategiskt levererade världen över för att möjliggöra innovation och kommersiell framgång.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material 2,3,4-Trihydroxyacetophenone
CAS 528-21-2
Molekylformel C8H8O4
Kvantitet 10 g
Synonym 1-(2,3,4-Trihydroxyphenyl)-ethanone, 2',3',4'-Trihydroxy-acetophenone, Gallacetophenone, 1-(2,3,4-Trihydroxyphenyl)ethanone, 2',3',4'-Trihydroxyacetophenone, 4-Acetylpyrogallol, Alizarin Yellow C, Alizarine Yellow C, C.I. 57000, NSC 66553
InChI formel InChI=1S/C8H8O4/c1-4(9)5-2-3-6(10)8(12)7(5)11/h2-3,10-12H,1H3
LEDER CC(=O)c1ccc(O)c(O)c1O
IUPAC-namn 1-(2,3,4-trihydroxyfenyl)etanon
Molekylvikt (g/mol) 168.15
Formel vikt 168.0423
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.