Learn More
2,3,4-Trimethyl-3-pentanol, 98%, Thermo Scientific™
Product Code. 11449706
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11449706 5 g 5g
11459706 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11449706 Supplier Thermo Scientific Alfa Aesar Supplier No. B20236.06
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3054-92-0
Molecular Formula C8H18O
Molecular Weight (g/mol) 130.231
MDL Number MFCD00048319
InChI Key PLSMHHUFDLYURK-UHFFFAOYSA-N
Synonym 2,3,4-trimethyl-3-pentanol, 1,1-diisopropylethanol, 3-pentanol, 2,3,4-trimethyl, diisopropylmethylcarbinol, unii-e7oyt9fj8f, e7oyt9fj8f, fema no. 3903, 2,3,4-trimethyl-3-pentanol fhfi, acmc-1cqn4, 2,3,4-trimethyl-pentan-3-ol
PubChem CID 520484
IUPAC Name 2,3,4-trimethylpentan-3-ol
SMILES CC(C)C(C)(C(C)C)O
Specifications
Quantity 5 g
Formula Weight 130.23
Percent Purity 98%
Chemical Name or Material 2,3,4-Trimethyl-3-pentanol