missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 2,3:4,6-Di-O-isopropylidene-alpha-L-sorbofuranose, 98%

Catalog Number p-7043218
Click to view available options
Quantity:
5 g
10 g
25 g
This item is not returnable. View return policy
14131-84-1
C12H20O6
260.286
MFCD00134206
JWWCLCNPTZHVLF-WVHJSVDISA-N
2,3:5,6-bis-o-1-methylethylidene-, a-d-mannofuranose, 3ar,4s,6r-6-2,2-dimethyl-1,3-dioxolan-4-yl-2,2-dimethyl-tetrahydrofuro 3,4-d 1,3 dioxol-4-ol
45039045
(3aR,4S,6R)-6-(2,2-dimethyl-1,3-dioxolan-4-yl)-2,2-dimethyl-3a,4,6,6a-tetrahydrofuro[3,4-d][1,3]dioxol-4-ol
CC1(OCC(O1)C2C3C(C(O2)O)OC(O3)(C)C)C
This item is not returnable. View return policy
14131-84-1
C12H20O6
260.286
MFCD00134206
JWWCLCNPTZHVLF-WVHJSVDISA-N
2,3:5,6-bis-o-1-methylethylidene-, a-d-mannofuranose, 3ar,4s,6r-6-2,2-dimethyl-1,3-dioxolan-4-yl-2,2-dimethyl-tetrahydrofuro 3,4-d 1,3 dioxol-4-ol
45039045
(3aR,4S,6R)-6-(2,2-dimethyl-1,3-dioxolan-4-yl)-2,2-dimethyl-3a,4,6,6a-tetrahydrofuro[3,4-d][1,3]dioxol-4-ol
CC1(OCC(O1)C2C3C(C(O2)O)OC(O3)(C)C)C

2,3:5,6-Di-O-isopropylidene-alpha-D-mannofuranose is used in the syntheses of ovalicin1 and of the sugar core of hikizimycin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3:5,6-Di-O-isopropylidene-alpha-D-mannofuranose is used in the syntheses of ovalicin1 and of the sugar core of hikizimycin.

Solubility
Soluble in acetone, methanol, water.

Notes
Hygroscopic. Protect from heat. Store under dry inert gas. Incompatible with moisture and oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 125°C to 126°C (decomposition)
Color White
Quantity 5 g
Beilstein 84382
Sensitivity Hygroscopic
Solubility Information Soluble in acetone,methanol,water.
Formula Weight 260.28
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2,3:5,6-Di-O-isopropylidene-alpha-D-mannofuranose

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'