Läs mer
2,3,5,6-Tetradeuterio-4-methoxyaniline, TRC
Produktkod. 30417760
Change view
Kvantitet:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30417760 5 mg
30423528 10 mg
30425460 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30417760 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT1219795MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1219798-55-6
Molekylformel C7 D4 H5 N O
Molekylvikt (g/mol) 127.177
Synonym Benzen-2,3,5,6-d4-amine, 4-methoxy- (ACI), 4-Methoxybenzen-2,3,5,6-d4-amine (ACI), 2,3,5,6-Tetradeuterio-4-methoxyaniline, 4-Methoxyaniline-2,3,5,6-d4, 4-Methoxyaniline-2,3,5,6-D4, p-Anisidine-d4
IUPAC-namn 2,3,5,6-tetradeuterio-4-methoxyaniline
LEDER [2H]c1c([2H])c(OC)c([2H])c([2H])c1N
Specifikationer
Kemiskt namn eller material 2,3,5,6-Tetradeuterio-4-methoxyaniline
Kvantitet 5 mg
InChI formel InChI=1S/C7H9NO/c1-9-7-4-2-6(8)3-5-7/h2-5H,8H2,1H3/i2D,3D,4D,5D
Formel vikt 127.094