Läs mer
2,3,5,6-Tetraphenylpyrazine, TRC
Produktkod. 30411297
Change view
Kvantitet:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30411297 250 mg
30420632 500 mg
30418496 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30411297 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT204561250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 642-04-6
Molekylformel C28H20N2
Molekylvikt (g/mol) 384.47
Synonym 2,3,5,6-Tetraphenyl-1,4-pyrazine, Amaron, NSC 123074, Tetraphenylpyrazine
IUPAC-namn 2,3,5,6-tetraphenylpyrazine
LEDER c1ccc(cc1)c2nc(c3ccccc3)c(nc2c4ccccc4)c5ccccc5
Specifikationer
Kemiskt namn eller material 2,3,5,6-Tetraphenylpyrazine
Kvantitet 250 mg
InChI formel InChI=1S/C28H20N2/c1-5-13-21(14-6-1)25-26(22-15-7-2-8-16-22)30-28(24-19-11-4-12-20-24)27(29-25)23-17-9-3-10-18-23/h1-20H
Formel vikt 384.1626