Läs mer
2,3,5-Trichloroaniline, TRC
Produktkod. 30180017
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30180017 100 mg
30180122 10 mg
30180379 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30180017 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT899565100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 18487-39-3
Molekylformel C6H4NCl3
Molekylvikt (g/mol) 196.46
Synonym 2,3,5-Trichloroaniline, Aniline, 2,3,5-trichloro- (6CI,8CI), 2,3,5-Trichlorobenzenamine
IUPAC-namn 2,3,5-trichloroaniline
LEDER Nc1cc(Cl)cc(Cl)c1Cl
Specifikationer
Kemiskt namn eller material 2,3,5-Trichloroaniline
Kvantitet 100 mg
InChI formel InChI=1S/C6H4Cl3N/c7-3-1-4(8)6(9)5(10)2-3/h1-2H,10H2
Formel vikt 194.9409