Läs mer
2,3,5-Trichlorobenzoic Acid, TRC
Produktkod. 30454869
Change view
Kvantitet:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30454869 5 g
30451897 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30454869 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT7743455G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 50-73-7
Molekylformel C7 H3 Cl3 O2
Molekylvikt (g/mol) 225.46
Synonym Benzoic acid, 2,3,5-trichloro-, 2,3,5-Trichlorobenzoic acid
IUPAC-namn 2,3,5-trichlorobenzoic acid
LEDER OC(=O)c1cc(Cl)cc(Cl)c1Cl
Specifikationer
Kemiskt namn eller material 2,3,5-Trichlorobenzoic acid
Kvantitet 5 g
InChI formel InChI=1S/C7H3Cl3O2/c8-3-1-4(7(11)12)6(10)5(9)2-3/h1-2H,(H,11,12)
Formel vikt 223.9199