Learn More
2,3,5-Trimethylfuran, 99%, Thermo Scientific Chemicals
Catalog Number p-7077484
859.96 SEK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
1 g
5 g
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
10504-04-8
C7H10O
110.16
MFCD03931179
NJXZFRUNHWKHEC-UHFFFAOYSA-N
furan, 2,3,5-trimethyl, 2,3,5-trimethyl-furan, furan,2,3,5-trimethyl
517859
2,3,5-trimethylfuran
CC1=CC(C)=C(C)O1
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
10504-04-8
C7H10O
110.16
MFCD03931179
NJXZFRUNHWKHEC-UHFFFAOYSA-N
furan, 2,3,5-trimethyl, 2,3,5-trimethyl-furan, furan,2,3,5-trimethyl
517859
2,3,5-trimethylfuran
CC1=CC(C)=C(C)O1
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Density 0.8900g/mL
Boiling Point 121.0°C to 122.0°C
Flash Point 20°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Refractive Index 1.449
Quantity 1 g
Specific Gravity 0.89
Formula Weight 110.16
Percent Purity 99%
Chemical Name or Material 2,3,5-Trimethylfuran
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2,3,5-Trimethylfuran, 99%
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.