Läs mer
2,3,5-Trimethylphenyl Trifluoromethanesulfonate, TRC
Produktkod. 30425966
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30425966 100 mg
1 options
Produktkod. 30425966 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT302760100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2,3,5-Trimethylphenyl Trifluoromethanesulfonate
CAS 209684-76-4
Molekylformel C10H11F3O3S
Kvantitet 100 mg
Synonym Trifluoromethanesulfonic Acid 2,3,5-Trimethylphenyl Ester, 1,1,1-Trifluoromethansulfonic Acid 2,3,5-Trimethylphenyl Ester
InChI formel InChI=1S/C10H11F3O3S/c1-6-4-7(2)8(3)9(5-6)16-17(14,15)10(11,12)13/h4-5H,1-3H3
LEDER O=S(OC1=C(C)C(C)=CC(C)=C1)(C(F)(F)F)=O
IUPAC-namn 2,3,5-trimethylphenyl trifluoromethanesulfonate
Molekylvikt (g/mol) 268.25
Formel vikt 268.04