Learn More
Alfa Aesar™ 2-(3-Aminophenyl)-4-(4-methoxyphenyl)thiazole, 97%
Brand:  Alfa Aesar™ H65954.03
Additional Details : Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15492669
  • 2420.01 SEK / 1g
Estimated Shipment date
from Supplier 30-07-2018
Add to basket
Specifications
Chemical Identifiers
2-(3-Aminophenyl)-4-(4-methoxyphenyl)thiazole
MFCD06739778
Specifications
2-(3-Aminophenyl)-4-(4-methoxyphenyl)thiazole
Yellow
97%
Air Sensitive
282.36
Solid
1g
MFCD06739778