Learn More
Alfa Aesar™ 2-(3-Benzoylphenyl)-N-(2,5-dichlorophenyl)propionamide, 95%
Brand:  Alfa Aesar™ H65912.03
Additional Details : Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15489996
  • 2362.17 SEK / 1g
Estimated Shipment date
from Supplier 12-03-2018
Add to basket
Specifications
Chemical Identifiers
2-(3-Benzoylphenyl)-N-(2,5-dichlorophenyl)propionamide
398.28
MFCD03838515
Specifications
2-(3-Benzoylphenyl)-N-(2,5-dichlorophenyl)propionamide
95%
MFCD03838515
Solid
398.28
1g