missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(3-Benzoylphenyl)-N-(2-ethoxyphenyl)propionamide, 95%

Brand:  Alfa Aesar™ H65092.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15489366

  • 2603.20 SEK / 1g


Estimated Shipment date
from Supplier 03-08-2018
Add to basket

Specifications

Chemical Identifiers

2-(3-Benzoylphenyl)-N-(2-ethoxyphenyl)propionamide
MFCD28098748

Specifications

2-(3-Benzoylphenyl)-N-(2-ethoxyphenyl)propionamide
Solid
MFCD28098748
373.44
95%
1g
Videos
SDS