missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(3-Benzoylphenyl)-N-(2-methylbenzyl)propionamide, 95%

Brand:  Alfa Aesar™ H65337.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15499566

  • 2362.17 SEK / 1g


Estimated Shipment date
from Supplier 08-03-2018
Add to basket

Specifications

Chemical Identifiers

2-(3-Benzoylphenyl)-N-(2-methylbenzyl)propionamide
357.44

Specifications

2-(3-Benzoylphenyl)-N-(2-methylbenzyl)propionamide
95%
1g
Solid
357.44
Videos
SDS