Learn More
Alfa Aesar™ 2-(3-Benzoylphenyl)-N-(3-fluoro-2-methylphenyl)propionamide, 95%
Brand:  Alfa Aesar™ H65799.03
Additional Details : Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15449906
  • 2603.20 SEK / 1g
Estimated Shipment date
from Supplier 15-11-2018
Add to basket
Specifications
Chemical Identifiers
MFCD28098825
Specifications
2-(3-Benzoylphenyl)-N-(3-fluoro-2-methylphenyl)propionamide
MFCD28098825
Solid
1g
361.41
95%