missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(3-Benzoylphenyl)-N-(3-methylphenyl)propionamide, 95%

Brand:  Alfa Aesar™ H65346.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15449576

  • 2603.20 SEK / 1g


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

2-(3-Benzoylphenyl)-N-(3-methylphenyl)propionamide
343.32
MFCD03838519
2-(3-Benzoylphenyl)-N-(m-tolyl)propionamide

Specifications

2-(3-Benzoylphenyl)-N-(3-methylphenyl)propionamide
95%
MFCD03838519
1g
Solid
343.32
2-(3-Benzoylphenyl)-N-(m-tolyl)propionamide
Videos
SDS