Learn More
Alfa Aesar™ 2-(3-Benzoylphenyl)-N-(4-bromo-2-chlorophenyl)propionamide, 95%
Brand:  Alfa Aesar™ H65889.03
Additional Details : Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15449976
  • 2603.20 SEK / 1g
Estimated Shipment date
from Supplier 02-07-2018
Add to basket
Specifications
Chemical Identifiers
2-(3-Benzoylphenyl)-N-(4-bromo-2-chlorophenyl)propionamide
442.73
MFCD28098836
Specifications
2-(3-Benzoylphenyl)-N-(4-bromo-2-chlorophenyl)propionamide
95%
MFCD28098836
Solid
442.73
1g