Learn More
2-(3-Bromophenyl)propan-2-ol, 97%, Thermo Scientific™
Catalog Number p-4812640
Click to view available options
Quantity:
1 g
5 g
30951-66-7
C9H11BrO
215.09
ZRFMJMFYMQAUDO-UHFFFAOYSA-N
2-3-bromophenyl propan-2-ol, 2-3-bromophenyl-2-propanol, acmc-209hjh, 2-3-bromophenyl-propan-2-ol, 2-3-bromo-phenyl-propan-2-ol, 3-alpha-hydroxyisopropyl-1-bromobenzene
15072383
2-(3-bromophenyl)propan-2-ol
CC(C)(C1=CC(=CC=C1)Br)O
30951-66-7
C9H11BrO
215.09
ZRFMJMFYMQAUDO-UHFFFAOYSA-N
2-3-bromophenyl propan-2-ol, 2-3-bromophenyl-2-propanol, acmc-209hjh, 2-3-bromophenyl-propan-2-ol, 2-3-bromo-phenyl-propan-2-ol, 3-alpha-hydroxyisopropyl-1-bromobenzene
15072383
2-(3-bromophenyl)propan-2-ol
CC(C)(C1=CC(=CC=C1)Br)O
Boiling Point 100°C (3.0 Torr)
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 215.09
Percent Purity 97%
Chemical Name or Material 2-(3-Bromophenyl)propan-2-ol
compliance-icons
Product Identifier
  • 2-(3-Bromophenyl)propan-2-ol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.