Learn More
2-(3-Bromophenyl)pyridine, 97%, Acros Organics™
Brand:  Acros Organics 457560010
Additional Details : CAS Number : 4373-60-8 Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15462182
  • 955.44 SEK / 1g
Estimated Shipment date
from Supplier 27-06-2018
Add to basket
Specifications
Chemical Identifiers
Authentic
4373-60-8
97%
234.1
C11H8BrN
2-(3-Bromophenyl)pyridine
Specifications
2-(3-Bromophenyl)pyridine
Authentic
97%
234.1
4373-60-8
1g
C11H8BrN
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • 2-(3-Bromophenyl)pyridine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.