missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(3-Bromophenyl)thiazolidine-4-carboxylic acid, 97%

Brand:  Alfa Aesar™ H62875.06

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15467946

  • 10374.25 SEK / 5g


Estimated Shipment date
from Supplier 03-07-2018
Add to basket

Specifications

Chemical Identifiers

2-(3-Bromophenyl)thiazolidine-4-carboxylic acid
288.16
MFCD00176218

Specifications

2-(3-Bromophenyl)thiazolidine-4-carboxylic acid
Solid
288.16
5g
97%
MFCD00176218
Videos
SDS