missing translation for 'onlineSavingsMsg'
Learn More

2-(3-Bromopropoxy)tetrahydro-2H-pyran, 98%, stabilized with potassium carbonate, Thermo Scientific Chemicals

Catalog Number p-6827178
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
33821-94-2
C8H15BrO2
223.1
HJNHUFQGDJLQRS-UHFFFAOYSA-N
2-3-bromopropoxy tetrahydro-2h-pyran, 2-3-bromopropoxy oxane, 2-3-bromopropoxy-tetrahydro-2h-pyran, 2-3-bromopropoxy tetrahydropyran, 1-bromo-3-tetrahydropyranyloxypropane, 2-3-bromo-propoxy-tetrahydro-pyran, 2h-pyran, 2-3-bromopropoxy tetrahydro, acmc-1cjr6, 1-bromo-3-2-oxanyloxy propane
2777988
2-(3-bromopropoxy)oxane
C1CCOC(C1)OCCCBr
This item is not returnable. View return policy
33821-94-2
C8H15BrO2
223.1
HJNHUFQGDJLQRS-UHFFFAOYSA-N
2-3-bromopropoxy tetrahydro-2h-pyran, 2-3-bromopropoxy oxane, 2-3-bromopropoxy-tetrahydro-2h-pyran, 2-3-bromopropoxy tetrahydropyran, 1-bromo-3-tetrahydropyranyloxypropane, 2-3-bromo-propoxy-tetrahydro-pyran, 2h-pyran, 2-3-bromopropoxy tetrahydro, acmc-1cjr6, 1-bromo-3-2-oxanyloxy propane
2777988
2-(3-bromopropoxy)oxane
C1CCOC(C1)OCCCBr

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.3140g/mL
Boiling Point 64.5°C (0.8 mmHg)
Flash Point 106°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Specific Gravity 1.314
Formula Weight 223.1
Percent Purity 98%
Chemical Name or Material 2-(3-Bromopropoxy)tetrahydro-2H-pyran, Stabilized with potassium carbonate
compliance-icons
Product Identifier
  • 2-(3-Bromopropoxy)tetrahydro-2H-pyran, stabilized
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'