Läs mer
2-(3-Chlorophenyl)-2-methylpropan-1-amine, TRC
Produktkod. 30351096
Change view
Kvantitet:
50 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30351096 100 mg
30366872 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30351096 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC428785100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-(3-Chlorophenyl)-2-methylpropan-1-amine
CAS 92015-24-2
Molekylformel C10H14ClN
Kvantitet 100 mg
Synonym 3-Chloro-beta,beta-dimethylbenzeneethanamine, 2-(3-Chloro-phenyl)-2-methyl-propylamine, m-Chloro-beta,beta-dimethyl-phenethylamine
InChI formel InChI=1S/C10H14ClN/c1-10(2,7-12)8-4-3-5-9(11)6-8/h3-6H,7,12H2,1-2H3
LEDER CC(C)(CN)c1cccc(Cl)c1
IUPAC-namn 2-(3-chlorophenyl)-2-methylpropan-1-amine
Molekylvikt (g/mol) 183.68
Formel vikt 183.0815