Läs mer
2-(3-Chloropropyl)-2-(4-fluorophenyl)-1,3-dioxolane (~90%), TRC
Produktkod. 30350897
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30350897 100 mg
1 options
Produktkod. 30350897 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC390550100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-(3-Chloropropyl)-2-(4-fluorophenyl)-1,3-dioxolane
CAS 3308-94-9
Molekylformel C12 H14 Cl F O2
Kvantitet 100 mg
Synonym 1,1-(Ethylenedioxy)-4-chloro-1-(4-fluorophenyl)butane, 1-(4-Fluorophenyl)-4-chlorobutan-1-one Ethylene Ketal, 1-Chloro-4,4-(ethylenedioxy)-4-(4-fluorophenyl)butane, 2-(3-Chloropropyl)-2-(4-fluorophenyl)dioxolane, 2-(3-Chloropropyl)-2-(4'-fluorophenyl)-1,3-dioxolane, 2-(3-Chloropropyl)-2-(p-fluorophenyl)-1,3-dioxolane, 2-(3-Chloropropyl)-2-(p-fluorophenyl)dioxolane, 2-(4-Fluorophenyl)-2-(3-chloropropyl)-1,3-dioxolane, 3-Chloropropyl 4-Fluorophenyl Ketone Ethylene Ketal, 3-p-Fluorobenzoylpropyl Chloride Ethylene Ketal, 4,4-(Ethylenedioxy)-4-(4-fluorophenyl)butyl Chloride, 4-(4-Fluorophenyl)-4,4-ethylenedioxy-1-chlorobutane, 4-(p-Fluorophenyl)-4,4-ethylenedioxy-1-chlorobutane, 4-Chloro-1,1-ethylenedioxy-1-(4-fluorophenyl)butane, 4-Chloro-1-(p-fluorophenyl)-1-butanone Cyclic Ethylene Acetal, 4-Chloro-4'-fluorobutyrophenone Cyclic Ethylene Ketal, 4-Chloro-p-fluorobutyrophenone Cyclic Ethylene Acetal, 4'-Fluoro-4-chlorobutyrophenone Ethylene Ketal
InChI formel InChI=1S/C12H14ClFO2/c13-7-1-6-12(15-8-9-16-12)10-2-4-11(14)5-3-10/h2-5H,1,6-9H2
LEDER Fc1ccc(cc1)C2(CCCCl)OCCO2
IUPAC-namn 2-(3-chloropropyl)-2-(4-fluorophenyl)-1,3-dioxolane
Molekylvikt (g/mol) 244.69
Formel vikt 244.0666