Läs mer
2,3-Dehydro Simvastatin Acid Sodium Salt, TRC
Produktkod. 30364759
Change view
Kvantitet:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30364759 5 mg
30356505 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30364759 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD2305505MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 393825-04-2
Molekylformel C25 H37 O5 . Na
Molekylvikt (g/mol) 440.55
IUPAC-namn sodium;(E,5R)-7-[(1S,2S,6R,8S,8aR)-8-(2,2-dimethylbutanoyloxy)-2,6-dimethyl-1,2,6,7,8,8a-hexahydronaphthalen-1-yl]-5-hydroxyhept-2-enoate
LEDER [Na+].CCC(C)(C)C(=O)O[C@H]1C[C@@H](C)C=C2C=C[C@H](C)[C@H](CC[C@@H](O)C\C=C\C(=O)[O-])[C@@H]12
Specifikationer
Kemiskt namn eller material 2,3-Dehydro Simvastatin Acid Sodium Salt
Kvantitet 5 mg
InChI formel InChI=1S/C25H38O5.Na/c1-6-25(4,5)24(29)30-21-15-16(2)14-18-11-10-17(3)20(23(18)21)13-12-19(26)8-7-9-22(27)28;/h7,9-11,14,16-17,19-21,23,26H,6,8,12-13,15H2,1-5H3,(H,27,28);/q;+1/p-1/b9-7+;/t16-,17-,19-,20-,21-,23-;/m0./s1
Formel vikt 440.2539