Läs mer
2’,3’-Di-O-acetyl-5’-deoxy-5-fluorocytidine, TRC
Produktkod. 30132777
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30132777 250 mg
30005750 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30132777 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD322500250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 161599-46-8
Molekylformel C13 H16 F N3 O6
Molekylvikt (g/mol) 329.28
Synonym 5'-Deoxy-5-fluorocytidine 2',3'-diacetate, 2',3'-Di-O-acetyl-5'-deoxy-5-fluorocytidine, 5'-Deoxy-2',3'-di-O-acetyl-5-fluorocytidine, Capecitabine Imp. C (EP)
IUPAC-namn [(2R,3R,4R,5R)-4-acetyloxy-5-(4-amino-5-fluoro-2-oxopyrimidin-1-yl)-2-methyloxolan-3-yl] acetate
LEDER C[C@H]1O[C@H]([C@H](OC(=O)C)[C@@H]1OC(=O)C)N2C=C(F)C(=NC2=O)N
Specifikationer
Kemiskt namn eller material 2',3'-Di-O-acetyl-5'-deoxy-5-fluorocytidine
Kvantitet 250 mg
InChI formel InChI=1S/C13H16FN3O6/c1-5-9(22-6(2)18)10(23-7(3)19)12(21-5)17-4-8(14)11(15)16-13(17)20/h4-5,9-10,12H,1-3H3,(H2,15,16,20)/t5-,9-,10-,12-/m1/s1
Formel vikt 329.1023