missing translation for 'onlineSavingsMsg'
Learn More

2,3-Diaminobenzoic acid, 95%, ACROS Organics™

Brand:  Acros Organics 457490250

Additional Details : CAS Number : 603-81-6 Weight : 0.02500kg


Quantity 25g

 View more versions of this product

Product Code. 15565976

  • 4278.00 SEK / 25g


Estimated Shipment
09-12-2020
Add to basket

Description

Description

Specifications

Chemical Identifiers

603-81-6
152.15
diaminobenzoic acid, benzoic acid, diamino, 3-carboxy-1,2-diaminobenzene, pubchem3145, acmc-209mjn, 2,3-diaminobenzonic acid, 2,3-diamino benzoic acid, ksc203g4l, pharmabridge p-2952, 2,3-diaminobenzoic acid
2,3-diaminobenzoic acid
C7H8N2O2
KKTUQAYCCLMNOA-UHFFFAOYSA-N
198069
C1=CC(=C(C(=C1)N)N)C(=O)O

Specifications

2,3-Diaminobenzoic acid
232.0°C to 234.5°C
603-81-6
C7H8N2O2
KKTUQAYCCLMNOA-UHFFFAOYSA-N
2,3-diaminobenzoic acid
198069
95%
Authentic
25g
94% min. (HPLC)
diaminobenzoic acid, benzoic acid, diamino, 3-carboxy-1,2-diaminobenzene, pubchem3145, acmc-209mjn, 2,3-diaminobenzonic acid, 2,3-diamino benzoic acid, ksc203g4l, pharmabridge p-2952, 2,3-diaminobenzoic acid
C1=CC(=C(C(=C1)N)N)C(=O)O
152.15
152.15
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2,3-Diaminobenzoic acid, 95%

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.

SDS
Documents