Läs mer
2,3-Diaminobenzoic Acid Methyl Ester Hydrochloride, TRC
Produktkod. 30351265
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30351265 500 mg
1 options
Produktkod. 30351265 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD416211500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2,3-Diaminobenzoic Acid Methyl Ester Hydrochloride
CAS 1189942-66-2
Molekylformel C8H11ClN2O2
Kvantitet 500 mg
Synonym Methyl 2,3-Diaminobenzoate Hydrochloride
InChI formel InChI=1S/C8H10N2O2.ClH/c1-12-8(11)5-3-2-4-6(9)7(5)10;/h2-4H,9-10H2,1H3;1H
LEDER Cl.COC(=O)c1cccc(N)c1N
Molekylvikt (g/mol) 202.64
Formel vikt 202.0509