Läs mer
2,3-Dibromo-3-(p-methoxyl)phenyl Propionic Acid, TRC
Produktkod. 30377936
Change view
Kvantitet:
25 g
Produktkod. Kvantitet
30377936 25 g
1 options
Produktkod. 30377936 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD42545525G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2,3-Dibromo-3-(p-methoxyl)phenyl Propionic Acid
CAS 77820-32-7
Molekylformel C10 H10 Br2 O3
Kvantitet 25 g
Synonym Benzenepropanoic acid, α,β-dibromo-4-methoxy-, Hydrocinnamic acid, α,β-dibromo-p-methoxy- (6CI,7CI), α,β-Dibromo-4-methoxybenzenepropanoic acid, 2,3-Dibromo-3-(4-methoxyphenyl)propanoic acid, 2,3-Dibromo-p-methoxyhydrocinnamic acid
InChI formel InChI=1S/C10H10Br2O3/c1-15-7-4-2-6(3-5-7)8(11)9(12)10(13)14/h2-5,8-9H,1H3,(H,13,14)
LEDER COc1ccc(cc1)C(Br)C(Br)C(=O)O
IUPAC-namn 2,3-dibromo-3-(4-methoxyphenyl)propanoic acid
Molekylvikt (g/mol) 337.99
Formel vikt 335.8997