missing translation for 'onlineSavingsMsg'
Läs mer

2,3-Dichloro-5-hydroxybenzaldehyde, TRC

Product Code. 30377682
Change view
Click to view available options
Kvantitet:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30377682 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30377682 Supplier Toronto Research Chemicals Supplier No. TRCD4349251G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material 2,3-Dichloro-5-hydroxybenzaldehyde
CAS 1806281-75-3
Molekylformel C7 H4 Cl2 O2
Kvantitet 1 g
Synonym Benzaldehyde, 2,3-dichloro-5-hydroxy- (ACI), 2,3-Dichloro-5-hydroxybenzaldehyde (ACI)
InChI formel InChI=1S/C7H4Cl2O2/c8-6-2-5(11)1-4(3-10)7(6)9/h1-3,11H
LEDER ClC1=C(C=O)C=C(O)C=C1Cl
IUPAC-namn 2,3-dichloro-5-hydroxybenzaldehyde
Molekylvikt (g/mol) 191.01
Formel vikt 189.9588
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.