missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichloro-5-(trifluoromethyl)pyridine, 97%, Thermo Scientific Chemicals

Catalog Number p-3757629
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
69045-84-7
C6H2Cl2F3N
215.98
MFCD00042243
ABNQGNFVSFKJGI-UHFFFAOYSA-N
2,3-dichloro-5-trifluoromethyl pyridine, unii-5f2c8lx8ie, 2,3-dichloro-5-trifluoro methylpyridine, 2,3,5-dctf, 5f2c8lx8ie, 2,3-dichloro-5-trifluoromethyl-pyridine, pyridine, 2,3-dichloro-5-trifluoromethyl, pubchem9237
112234
2,3-dichloro-5-(trifluoromethyl)pyridine
FC(F)(F)C1=CN=C(Cl)C(Cl)=C1
This item is not returnable. View return policy
69045-84-7
C6H2Cl2F3N
215.98
MFCD00042243
ABNQGNFVSFKJGI-UHFFFAOYSA-N
2,3-dichloro-5-trifluoromethyl pyridine, unii-5f2c8lx8ie, 2,3-dichloro-5-trifluoro methylpyridine, 2,3,5-dctf, 5f2c8lx8ie, 2,3-dichloro-5-trifluoromethyl-pyridine, pyridine, 2,3-dichloro-5-trifluoromethyl, pubchem9237
112234
2,3-dichloro-5-(trifluoromethyl)pyridine
FC(F)(F)C1=CN=C(Cl)C(Cl)=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 8.0°C to 9.0°C
Color Colorless to Yellow
Density 1.5490g/mL
Boiling Point 176.0°C
Flash Point 79°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4745 to 1.4765
Quantity 25 g
Specific Gravity 1.549
Formula Weight 216
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2, 3-Dichloro-5-(trifluoromethyl)pyridine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2,3-Dichloro-5-(trifluoromethyl)pyridine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'