Learn More
2,3-Dichloroanisole, 97%, Thermo Scientific Chemicals
Catalog Number p-7082025
Click to view available options
Quantity:
5 g
50 g
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1984-59-4
C7H6Cl2O
177.02
MFCD00000545
HFEASCCDHUVYKU-UHFFFAOYSA-N
2,3-dichloroanisole, benzene, 1,2-dichloro-3-methoxy, 2,3-dichloromethoxybenzene, 2,3-dichloranisol, dichloroanisole 2,3-, 1,2-dichloro-3-methoxy-benzene, dichloroanisol, 2,3-dichloro-anisol, pubchem3680, 2,3-dichloro anisole
16126
1,2-dichloro-3-methoxybenzene
COC1=CC=CC(Cl)=C1Cl
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1984-59-4
C7H6Cl2O
177.02
MFCD00000545
HFEASCCDHUVYKU-UHFFFAOYSA-N
2,3-dichloroanisole, benzene, 1,2-dichloro-3-methoxy, 2,3-dichloromethoxybenzene, 2,3-dichloranisol, dichloroanisole 2,3-, 1,2-dichloro-3-methoxy-benzene, dichloroanisol, 2,3-dichloro-anisol, pubchem3680, 2,3-dichloro anisole
16126
1,2-dichloro-3-methoxybenzene
COC1=CC=CC(Cl)=C1Cl
Synthesis Material Intermediates. The product has been used to develop an analytical method for the determination of biogenic halogenated anisoles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Synthesis Material Intermediates. The product has been used to develop an analytical method for the determination of biogenic halogenated anisoles.
Solubility
Soluble in water (86.9 mg/L).
Notes
Store in cool, dry conditions in well sealed containers. Store away from strong oxidizing agents, strong acids. Keep container tightly closed.
TRUSTED_SUSTAINABILITY
Melting Point 31°C to 33°C
Boiling Point 115°C to 116°C (12 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
Beilstein 1909166
Solubility Information Soluble in water (86.9mg/L).
Formula Weight 177.03
Percent Purity 97%
Chemical Name or Material 2,3-Dichloroanisole