missing translation for 'onlineSavingsMsg'
Learn More

2,3-Difluoro-4-methylaniline, 97%, Thermo Scientific™

Catalog Number p-7029284
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
886503-79-3
C7H7F2N
143.14
MFCD06660321
HMDHQYKNBZEFTH-UHFFFAOYSA-N
2,3-difluoro-4-methylphenylamine, benzenamine,2,3-difluoro-4-methyl, 4-amino-2,3-difluoro-toluene
17750814
2,3-difluoro-4-methylaniline
CC1=CC=C(N)C(F)=C1F
This item is not returnable. View return policy
886503-79-3
C7H7F2N
143.14
MFCD06660321
HMDHQYKNBZEFTH-UHFFFAOYSA-N
2,3-difluoro-4-methylphenylamine, benzenamine,2,3-difluoro-4-methyl, 4-amino-2,3-difluoro-toluene
17750814
2,3-difluoro-4-methylaniline
CC1=CC=C(N)C(F)=C1F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Formula Weight 143.14
Percent Purity 97%
Chemical Name or Material 2,3-Difluoro-4-methylaniline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'