missing translation for 'onlineSavingsMsg'
Learn More

2,3-Difluoro-6-methylpyridine, 98%, Thermo Scientific Chemicals

Catalog Number p-7055835
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
1227579-04-5
C6H5F2N
129.11
MFCD16610828
HGBWAUANMSZOIJ-UHFFFAOYSA-N
2,3-difluoro-6-methyl-pyridine
70680696
2,3-difluoro-6-methylpyridine
CC1=NC(=C(C=C1)F)F
This item is not returnable. View return policy
1227579-04-5
C6H5F2N
129.11
MFCD16610828
HGBWAUANMSZOIJ-UHFFFAOYSA-N
2,3-difluoro-6-methyl-pyridine
70680696
2,3-difluoro-6-methylpyridine
CC1=NC(=C(C=C1)F)F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Quantity 250 mg
Formula Weight 129.11
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2,3-Difluoro-6-methylpyridine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'