Learn More
2,3-Difluorobenzyl alcohol, 97%, Thermo Scientific™
Catalog Number p-7077226
Click to view available options
Quantity:
1 g
25 g
75853-18-8
C7H6F2O
144.12
JSFGDUIJQWWBGY-UHFFFAOYSA-N
2,3-difluorobenzyl alcohol, 2,3-difluorophenyl methanol, 2,3-difluorobenzylalcohol, 2,3-difluorophenylalcohol, benzenemethanol, 2,3-difluoro, 2,3-difluorophenyl methan-1-ol, pubchem7358, 2,3-diflorobenzyl alcohol, 2,3difluorophenyl methanol, 2,3 difluorophenyl methanol
447153
CHEBI:41883
(2,3-difluorophenyl)methanol
C1=CC(=C(C(=C1)F)F)CO
75853-18-8
C7H6F2O
144.12
JSFGDUIJQWWBGY-UHFFFAOYSA-N
2,3-difluorobenzyl alcohol, 2,3-difluorophenyl methanol, 2,3-difluorobenzylalcohol, 2,3-difluorophenylalcohol, benzenemethanol, 2,3-difluoro, 2,3-difluorophenyl methan-1-ol, pubchem7358, 2,3-diflorobenzyl alcohol, 2,3difluorophenyl methanol, 2,3 difluorophenyl methanol
447153
CHEBI:41883
(2,3-difluorophenyl)methanol
C1=CC(=C(C(=C1)F)F)CO
Density 1.2800g/mL
Flash Point 93°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Linear Formula F2C6H3CH2OH
Refractive Index 1.4910 to 1.4930
Quantity 1 g
Specific Gravity 1.28
Formula Weight 144.12
Percent Purity 97%
Chemical Name or Material 2,3-Difluorobenzyl alcohol