missing translation for 'onlineSavingsMsg'
Läs mer

2,3-Difluorobenzyl alcohol, 97%, Thermo Scientific™

Produktkod. 11434110
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
5 g
25 g
Förpackningsstorlek:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
11434110 25 g 25g
11430057 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 11434110 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer B20980.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter

Denna artikel kan inte returneras. Se returpolicy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used to produce 1-bromomethyl-2,3-difluoro-benzene with reagent PBr3 and solvent toluene at the temperature of 100°C for 1 hour. 2,3-Difluorobenzyl alcohol may be used in chemical synthesis studies.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 75853-18-8
Molekylformel C7H6F2O
Molekylvikt (g/mol) 144.121
MDL-nummer MFCD00010312
InChI-nyckel JSFGDUIJQWWBGY-UHFFFAOYSA-N
PubChem CID 447153
ChEBI CHEBI:41883
IUPAC-namn (2,3-difluorophenyl)methanol
LEDER C1=CC(=C(C(=C1)F)F)CO

Specifikationer

Kemiskt namn eller material 2,3-Difluorobenzyl alcohol
Densitet 1.282
Flampunkt 93°C (199°F)
Brytningsindex 1.492
Kvantitet 25 g
Beilstein 7089243
Synonym 2,3-difluorobenzyl alcohol, 2,3-difluorophenyl methanol, 2,3-difluorobenzylalcohol, 2,3-difluorophenylalcohol, benzenemethanol, 2,3-difluoro, 2,3-difluorophenyl methan-1-ol, pubchem7358, 2,3-diflorobenzyl alcohol, 2,3difluorophenyl methanol, 2,3 difluorophenyl methanol
Löslighetsinformation Slightly soluble in water.
Formel vikt 144.12
Procent renhet 97%
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.