Läs mer
2,3-Dihydroxy-2-methylbutanoic Acid Sodium Salt, TRC
Produktkod. 30008762
Change view
Kvantitet:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30008762 50 mg
30010269 5 mg
30008915 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30008762 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD45432850MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2,3-Dihydroxy-2-methylbutanoic Acid Sodium Salt
Anteckningar om renhetsgrad HPLC
Molekylformel C5 H9 Na O4
Kvantitet 50 mg
InChI formel InChI=1S/C5H10O4.Na/c1-3(6)5(2,9)4(7)8;/h3,6,9H,1-2H3,(H,7,8);/q;+1/p-1
LEDER CC(O)C(C)(O)C(=O)O[Na]
IUPAC-namn (2,3-dihydroxy-2-methylbutanoyl)oxysodium
Molekylvikt (g/mol) 156.112
Formel vikt 156.04
Procent renhet >95%