Läs mer
2,3-Dihydroxy-2-methylbutanoic Acid Sodium Salt, TRC
Produktkod. 30008915
Change view
Kvantitet:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30008915 10 mg
30008762 50 mg
30010269 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30008915 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD45432810MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2839062-01-8
Specifikationer
Kemiskt namn eller material 2,3-dihydroxy-2-methylbutanoic acid
Anteckningar om renhetsgrad HPLC
Molekylformel C5 H10 O4 . Na
Kvantitet 10 mg
InChI formel InChI=1S/C5H10O4/c1-3(6)5(2,9)4(7)8/h3,6,9H,1-2H3,(H,7,8)
LEDER [Na].O=C(O)C(O)(C)C(O)C
IUPAC-namn 2,3-dihydroxy-2-methyl-butanoic acid
Molekylvikt (g/mol) 157.12
Formel vikt 157.05
Procent renhet >95%