Läs mer
2,3-Dihydroxy-4-oxopentanoic Acid, TRC
Produktkod. 30133969
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133969 250 mg
30012455 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30133969 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD453920250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 37520-06-2
Molekylformel C5H8O5
Molekylvikt (g/mol) 148.11
Synonym 2,3-Dihydroxy-4-ketovaleric Acid, 5-Deoxy-4-pentulosonic Acid
LEDER OC(C(C)=O)C(O)C(O)=O
Specifikationer
Kemiskt namn eller material 2,3-Dihydroxy-4-oxopentanoic Acid
Anteckningar om renhetsgrad HPLC
Kvantitet 250 mg
InChI formel InChI=1S/C5H8O5/c1-2(6)3(7)4(8)5(9)10/h3-4,7-8H,1H3,(H,9,10)
Formel vikt 148.0372
Procent renhet >95%