Läs mer
2,3-Dihydroxypropanoic Acid(20% in water), TRC
Produktkod. 30133152
Change view
Kvantitet:
2.5 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133152 5 g
30136648 25 g
30011914 2.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30133152 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4934535G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 473-81-4
Molekylformel C3H6O4
Molekylvikt (g/mol) 106.08
Synonym Glyceric Acid
IUPAC-namn 2,3-dihydroxypropanoic acid
LEDER OCC(O)C(=O)O
Specifikationer
Kemiskt namn eller material 2,3-Dihydroxypropanoic Acid
Kvantitet 5 g
InChI formel InChI=1S/C3H6O4/c4-1-2(5)3(6)7/h2,4-5H,1H2,(H,6,7)
Formel vikt 106.0266
Koncentration eller sammansättning (efter analyt eller komponenter) 2,3-Dihydroxypropanoic Acid 20%