Läs mer
2,3-Diketogulonic Acid Potassium Salt (Technical Grade), TRC
Produktkod. 30011345
Change view
Kvantitet:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30011345 10 mg
30008211 50 mg
30008290 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30011345 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD46792510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2062656-92-0
Molekylformel C6 H7 O7 . K
Molekylvikt (g/mol) 230.21
Synonym 2,3-Diketogulonic Acid Potassium Salt (Technical Grade), Potassium (4R,5S)-4,5,6-Trihydroxy-2,3-dioxohexanoate
IUPAC-namn potassium;(4R,5S)-4,5,6-trihydroxy-2,3-dioxo-hexanoate
LEDER [K+].OC[C@H](O)[C@@H](O)C(=O)C(=O)C(=O)[O-]
Specifikationer
Kemiskt namn eller material 2,3-Diketogulonic Acid Potassium Salt
Kvantitet 10 mg
InChI formel InChI=1S/C6H8O7.K/c7-1-2(8)3(9)4(10)5(11)6(12)13;/h2-3,7-9H,1H2,(H,12,13);/q;+1/p-1/t2-,3+;/m0./s1
Formel vikt 229.9829