Läs mer
2,3-Dimercaptosuccinic Acid, TRC
Produktkod. 30374222
Change view
Kvantitet:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30374222 5 g
30373165 10 g
30373640 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30374222 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4865955G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2418-14-6
Molekylformel C4 H6 O4 S2
Molekylvikt (g/mol) 182.22
Synonym 2,3-Dimercaptobutanedioic Acid, DMSA
IUPAC-namn 2,3-bis(sulfanyl)butanedioic acid
LEDER OC(=O)C(S)C(S)C(=O)O
Specifikationer
Kemiskt namn eller material 2,3-Dimercaptosuccinic acid
Kvantitet 5 g
InChI formel InChI=1S/C4H6O4S2/c5-3(6)1(9)2(10)4(7)8/h1-2,9-10H,(H,5,6)(H,7,8)
Formel vikt 181.9708